missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Gold Chloride Trihydrate, Certified ACS, Fisher Chemical™
Manufacturer: Fisher Chemical G541
Description
- Used as a gold catalyst or source of gold for organic reactions
- Intended for research use only. Not for human or animal diagnostic purposes.
Specifications
Tetrachloroauric Acid Trihydrate | |
0.2% max. | |
MFCD00149904 | |
393.83 | |
≥49% | |
3.9g/cm³ | |
0.1% max. | |
Yellow | |
1g |
16961-25-4 | |
HAuCl4 · 3 H2O | |
Gold(III) chloride trihydrate, Tetrachloroauric acid trihydrate, Hydrogren tetrachloroaurate(III), Tetrachloroauric(III) acid, Chloroauric acid trihydrate | |
Solid | |
Certified ACS | |
Pass Test | |
Glass Vial | |
254°C |
Chemical Identifiers
16961-25-4 | |
HAuCl4 · 3 H2O | |
MFCD00149904 | |
Gold(III) chloride trihydrate, Tetrachloroauric acid trihydrate, Hydrogren tetrachloroaurate(III), Tetrachloroauric(III) acid, Chloroauric acid trihydrate |
Safety and Handling

Recommended Storage : Refrigerate