Please login to your online account to display your discounted pricing

Tetrachloro-p-benzoquinone, 99%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  213565000

 View more versions of this product

Catalog No. AC213565000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (HPLC)
Physical Form Liquid
Formula Weight 245.88
Melting Point >290°C
Quantity 500g
Color Clear
Beilstein 07, 636
Merck Index 15, 2078
RTECS Number DK6825000
EINECS Number 204-274-4
TSCA TSCA
Fieser 01,125; 02,66; 03,46; 04,75
Chemical Name or Material Tetrachloro-p-benzoquinone
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C6Cl4O2
Linear Formula C6Cl4(=O)2
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
MDL Number MFCD00001594
CAS 118-75-2
Synonym 2, 3, 5
Packaging Plastic bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Ash 0.5% max.

Specifications

Chemical Name or Material Tetrachloro-p-benzoquinone
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C6Cl4O2
Linear Formula C6Cl4(=O)2
Formula Weight 245.88
MDL Number MFCD00001594
Synonym 2, 3, 5
Accessories

Warning

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents