Please login to your online account to display your discounted pricing

Tetrachloro-p-benzoquinone, 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  213561000

 View more versions of this product

Catalog No. AC213561000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (HPLC)
Formula Weight 245.88
Melting Point >290°C
Physical Form Liquid
Quantity 100g
Color Clear
TSCA TSCA
RTECS Number DK6825000
EINECS Number 204-274-4
Fieser 01,125; 02,66; 03,46; 04,75
Merck Index 15, 2078
Beilstein 07, 636
Synonym 2, 3, 5
Infrared Spectrum Authentic
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Ash 0.5% max.
Chemical Name or Material Tetrachloro-p-benzoquinone
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6Cl4O2
Linear Formula C6Cl4(=O)2
MDL Number MFCD00001594
CAS 118-75-2
Assay Percent Range 99%

Specifications

Chemical Name or Material Tetrachloro-p-benzoquinone
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C6Cl4O2
Linear Formula C6Cl4(=O)2
Formula Weight 245.88
MDL Number MFCD00001594
Synonym 2, 3, 5
Accessories

Warning

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents