Please login to your online account to display your discounted pricing

Tetrachloro-o-benzoquinone 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  137990050

 View more versions of this product

Catalog No. AC137990050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Fine Crystalline Powder
Formula Weight 245.88
Quantity 5g
Color Bordeaux
EINECS Number 219-424-4
TSCA TSCA
Melting Point 125° to 129°C
Beilstein 07, 602
Fieser 01,128; 04,76; 05,104; 07,355; 09,102; 10,85
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Tetrachloro-o-benzoquinone
Name Note 97%
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6Cl4O2
Linear Formula C6Cl4(=O)2
MDL Number MFCD00001646
CAS 2435-53-2
Synonym o-Chloranil
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

Specifications

Chemical Name or Material Tetrachloro-o-benzoquinone
Name Note 97%
CAS 2435-53-2
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6Cl4O2
Linear Formula C6Cl4(=O)2
Formula Weight 245.88
MDL Number MFCD00001646
Synonym o-Chloranil
Accessories

Warning

GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

SDS & Safety Documents