missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrachloro-o-benzoquinone, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 137990250

Catalog No. AC137990250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Tetrachloro-o-benzoquinone
2435-53-2
96% min. (GC)
C6Cl4O2
MFCD00001646
07, 602
o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil
VRGCYEIGVVTZCC-UHFFFAOYSA-N
3,4,5,6-tetrachlorocyclohexa-3,5-diene-1,2-dione
73252
97%
97%
Authentic
Glass bottle
C6Cl4(=O)2
25g
01,128; 04,76; 05,104; 07,355; 09,102; 10,85
Solubility in water: insoluble. Other solubilities: 10g/L chloroform (20°C)
ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl
245.86
245.88
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 219-424-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only