Please login to your online account to display your discounted pricing

Tetracarbonyldi-μ-chlorodirhodium(I), 97%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  197602500

 View more versions of this product

Catalog No. AC197602500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0% (ex Rh)
Physical Form Crystals, Crystalline Powder or Needles
Formula Weight 388.76
Color Red to brownish
Quantity 250mg
Merck Index 15, 8312
EINECS Number 238-540-6
UN Number 2811
Chemical Name or Material Tetracarbonyldi-μ-chlorodirhodium(I)
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00135610
Synonym Rhodium carbonyl chloride
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C4Cl2O4Rh2
Linear Formula [Rh(CO)2Cl]2
CAS 14523-22-9

Specifications

Chemical Name or Material Tetracarbonyldi-μ-chlorodirhodium(I)
Assay ≥96.0% (ex Rh)
Assay Percent Range 97%
Molecular Formula C4Cl2O4Rh2
Linear Formula [Rh(CO)2Cl]2
Formula Weight 388.76
MDL Number MFCD00135610
Synonym Rhodium carbonyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Toxic if swallowed.
Causes serious eye irritation.

SDS & Safety Documents