missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium Phosphate (White to Off-White Powder/HPLC Grade), Fisher Chemical™

Improve HPLC separation of acidic analytes by ion pairing.

Manufacturer:  Fisher Chemical O611725

 View more versions of this product

Catalog No. O611725




Description

Description

  • High purity
  • Application tested
  • Dissolves easily
Specifications

Specifications

0.02 AU max. at 214nm
Pass Test
White
25g
5574-97-0
C16H38NO4P
MFCD00064526
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
dihydrogen phosphate;tetrabutylazanium
2735142
Powder/Solid
HPLC
Tetrabutylammonium Phosphate
Glass Bottle
148°C
4 to 5
≥98.5 %
(CH3CH2CH2CH2)4NH2PO4
1-butanaminium, n,n,n-tributyl-, phosphate 1:1, dihydrogen phosphate; tetrabutylammonium, dihydrogen phosphate; tetrabutylazanium, tetra-n-butylammonium phosphate, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium dihydrogen phosphate, tetrabutylammonium dihydrogenphosphate, tetrabutylammonium ion dihydrogenphosphate ion, tetrabutylammonium phosphate, tetrabutylazanium dihydrogen phosphate
CCCC[N+](CCCC)(CCCC)CCCC.OP(=O)(O)[O-]
339.457
339.45
≥98.5%

Chemical Identifiers

5574-97-0
339.457
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
2735142
CCCC[N+](CCCC)(CCCC)CCCC.OP(=O)(O)[O-]
C16H38NO4P
MFCD00064526
1-butanaminium, n,n,n-tributyl-, phosphate 1:1, dihydrogen phosphate; tetrabutylammonium, dihydrogen phosphate; tetrabutylazanium, tetra-n-butylammonium phosphate, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium dihydrogen phosphate, tetrabutylammonium dihydrogenphosphate, tetrabutylammonium ion dihydrogenphosphate ion, tetrabutylammonium phosphate, tetrabutylazanium dihydrogen phosphate
dihydrogen phosphate;tetrabutylazanium
Safety and Handling

Safety and Handling

SDS Documents