missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium Perchlorate, Contains Max. 10% Water 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 420141000

 View more versions of this product

Catalog No. AC420141000


 

Specifications Description & Specifications

Specifications

≥98.5%
Crystalline Powder
White
Glass bottle
1923-70-2
(CH3(CH2)3)4NClO4
Tetrabutylammonium perchlorate
C16H36ClNO4
341.91
100g
290°C
MFCD00038722
Authentic
99%
Contains Max. 10% Water

Chemical Identifiers

1923-70-2
C16H36ClNO4
341.91
Tetrabutylammonium perchlorate, Contains Max. 10% Water
99%
(CH3(CH2)3)4NClO4
MFCD00038722
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause fire or explosion; strong oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep/Store away from clothing/other combustible materials.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 217-655-5

TSCA : TSCA