missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium iodide, 98%, ACROS Organics™

Manufacturer:  Acros Organics 161270250

 View more versions of this product

Catalog No. AC161270250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Phase transfer catalyst
Specifications

Specifications

Tetrabutylammonium iodide
Authentic
Glass bottle
141°C to 150°C
311-28-4
C16H36IN
05,646; 06,566; 07,355; 09,447; 11,501
DPKBAXPHAYBPRL-UHFFFAOYSA-M
tetrabutylazanium;iodide
67553
Crystalline Powder
1% max.
1% max.
White to Cream
25g
98%
CH3(CH2)3!4NI
tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
CCCC[N+](CCCC)(CCCC)CCCC.[I-]
369.375
369.36
≥97.5% (Argentometry)

Chemical Identifiers

311-28-4
369.375
tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
tetrabutylazanium;iodide
C16H36IN
DPKBAXPHAYBPRL-UHFFFAOYSA-M
67553
CCCC[N+](CCCC)(CCCC)CCCC.[I-]
SDS Documents