Please login to your online account to display your discounted pricing

Tetrabutylammonium Hydroxide, MP Biomedicals™


Quantity 25mL

Manufacturer: mp biomedicals  0215210425

 View more versions of this product

Catalog No. ICN15210425

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 0.98g/mL
Physical State Liquid
Quantity 25mL
Chemical Name or Material Tetrabutylammonium Hydroxide
CAS 2052-49-5
Physical Form Clear colorless liquid
Molecular Formula C16H37NO
Formula Weight 259.474g/mol
Recommended Storage Store at room temperature
EINECS Number 218-147-6

Specifications

Chemical Name or Material Tetrabutylammonium Hydroxide
CAS 2052-49-5
Physical Form Clear colorless liquid
Molecular Formula C16H37NO
Formula Weight 259.474g/mol
Accessories

Store at room temperature