missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 176610500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetrabutylammonium hydroxide | |
2052-49-5,7732-18-5 | |
0.9950g/mL | |
Plastic bottle | |
[CH3(CH2)3]4NOH | |
50 g | |
05,645; 11,500 | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
tetrabutylazanium;hydroxide | |
2723671 | |
38 to 42% (Total base) |
40 wt.% Solution in Water | |
White to Yellow | |
>100.0°C | |
C16H37NO | |
MFCD00009425 | |
04, II, 634 | |
0.995 | |
Solubility in water: soluble. | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
259.48 | |
259.46 | |
Crystals or Powder |
RUO – Research Use Only