missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrabutylammonium Hydroxide (37% in Methanol), TCI America™
$41.48 - $248.86
Chemical Identifiers
CAS | 2052-49-5 |
---|---|
Molecular Formula | C16H37NO |
Molecular Weight (g/mol) | 259.48 |
MDL Number | MFCD00009425 |
InChI Key | VDZOOKBUILJEDG-UHFFFAOYSA-M |
Synonym | tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 |
PubChem CID | 2723671 |
IUPAC Name | tetrabutylazanium hydroxide |
SMILES | [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T194825ML
|
TCI America
T194825ML |
25 mL |
Each for $41.48
|
|
|||||
T1948500ML
|
TCI America
T1948500ML |
500 mL |
Each for $248.86
|
|
|||||
Chemical Identifiers
2052-49-5 | |
259.48 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
2723671 | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC |
C16H37NO | |
MFCD00009425 | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
tetrabutylazanium hydroxide |
Specifications
2052-49-5 | |
C16H37NO | |
25 mL | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
259.48 | |
259.48 | |
Tetrabutylammonium Hydroxide (37% in Methanol) |
Yellow | |
MFCD00009425 | |
2924 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
tetrabutylazanium hydroxide | |
2723671 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0186
RTECSNumber : BS5425000
TSCA : Yes