missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrabutylammonium hydroxide, 1M solution in methanol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 212918000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetrabutylammonium hydroxide | |
2052-49-5,67-56-1 | |
-98.0°C | |
[CH3(CH2)3]4NOH | |
0.9100g/mL | |
Solubility in water: soluble. | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
259.48 | |
259.46 | |
1.3775 | |
800 mL |
1M Solutionin Methanol | |
65.0°C | |
C16H37NO | |
MFCD00009425 | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
tetrabutylazanium hydroxide | |
2723671 | |
Plastic bottle | |
0.91 | |
12°C |
RUO – Research Use Only