Please login to your online account to display your discounted pricing

Tetrabutylammonium hydrogen sulfate, 99%, for HPLC, ACROS Organics™


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific acros  420105000

 View more versions of this product

Catalog No. AC420105000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Purity Grade HPLC Grade
Physical Form Powder or Crystals
Formula Weight 339.52
Quantity 500g
Color Colorless to white
EINECS Number 251-068-5
TSCA TSCA
Absorbance (0.005M in water) (1 cm cell vs water)
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Tetrabutylammonium hydrogen sulfate
Absorbance Conditions (0.005M in water) (1 cm cell vs water)
Synonym TBAHS
Packaging Glass Bottle
Assay Percent Range 99%
Molecular Formula C16H35N·H2SO4
Linear Formula [CH3(CH2)3]4NHSO4
MDL Number MFCD00011637
CAS 32503-27-8

Specifications

Chemical Name or Material Tetrabutylammonium hydrogen sulfate
CAS 32503-27-8
Purity Grade HPLC Grade
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C16H35N·H2SO4
Linear Formula [CH3(CH2)3]4NHSO4
Formula Weight 339.52
MDL Number MFCD00011637
Synonym TBAHS
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

SDS & Safety Documents