Please login to your online account to display your discounted pricing

Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  201950050

 View more versions of this product

Catalog No. AC201950050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 261.46
Quantity 5mL
EINECS Number 207-057-2
TSCA TSCA
Density 0.8870g/mL
Fieser 04,477; 05,645; 07,353; 08,467; 09,444; 10,378; 11,499; 12,458; 14,293; 15,168; 17,363
Synonym TBAF
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides.
CAS 109-99-9
Packaging Glass bottle
Chemical Name or Material Tetrabutylammonium fluoride
Name Note 1M Solution in THF, Containing ca 5% Water
Molecular Formula C16H36FN
Linear Formula [CH3(CH2)3]4NF
MDL Number MFCD00011747
CAS 429-41-4
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co
CAS 7732-18-5

Specifications

Chemical Name or Material Tetrabutylammonium fluoride
Name Note 1M Solution in THF, Containing ca 5% Water
CAS 429-41-4
CAS 109-99-9
CAS 7732-18-5
Molecular Formula C16H36FN
Linear Formula [CH3(CH2)3]4NF
Formula Weight 261.46
MDL Number MFCD00011747
Synonym TBAF
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides.

SDS & Safety Documents