Please login to your online account to display your discounted pricing

Tetrabutylammonium cyanoborohydride, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365060250

 View more versions of this product

Catalog No. AC365060250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 282.32
Physical Form Crystalline Powder
Quantity 25g
Color White
Melting Point 142° to 149°C
Fieser 05,645; 11,499
Health Hazard 1 GHS Signal Word: Warning
Linear Formula [CH3(CH2)3]4NBH3CN
MDL Number MFCD00012036
Assay Percent Range 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Tetrabutylammonium cyanoborohydride
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C17 H39 B N2
CAS 43064-96-6
Name Note 95%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
In contact with water releases flammable gas.

Specifications

Chemical Name or Material Tetrabutylammonium cyanoborohydride
Name Note 95%
CAS 43064-96-6
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C17 H39 B N2
Linear Formula [CH3(CH2)3]4NBH3CN
Formula Weight 282.32
MDL Number MFCD00012036
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
In contact with water releases flammable gas.

SDS & Safety Documents