Please login to your online account to display your discounted pricing

Tetrabutylammonium chloride hydrate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrabutylammonium Chloride Hydrate
CAS 37451-68-6
Assay ≥97.5% (on dried basis)
Assay Percent Range 98%
Molecular Formula Cl·xH2O
Formula Weight 277.92
MDL Number MFCD00149970
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432061000

  SDS
thermo fisher scientific
AC432061000
Glass Bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 277.92
Assay ≥97.5% (on dried basis)
Physical Form Low Melting Solid
Chemical Name or Material Tetrabutylammonium Chloride Hydrate
Packaging Glass Bottle
Quantity 100g
Color Off-white
Water 10% Max. (K.F.)
Melting Point 37° to 41°C
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 37451-68-6
Assay Percent Range 98%
Molecular Formula Cl·xH2O
MDL Number MFCD00149970
Infrared Spectrum Authentic

Phase transfer catalyst

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.