missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium chloride, 95%, Tech., contains also bromide, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 174125000

Catalog No. AC174125000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
Specifications

Specifications

Tetrabutylammonium chloride
1112-67-0
94% min. (on dry substance) (Argentometry)
C16H36ClN
MFCD00011635
04, III, 292
tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
NHGXDBSUJJNIRV-UHFFFAOYSA-M
tetrabutylazanium;chloride
70681
277.92
Technical
Also contains bromide
Authentic
Glass bottle
[CH3(CH2)3]4NCl
500g
06,565; 07,353; 17,363
Solubility in water: soluble.
CCCC[N+](CCCC)(CCCC)CCCC.[Cl-]
277.92
CHEBI:51988
95%
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 214-195-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only