Learn More
Tetrabutylammonium chloride, 95%, Tech., contains also bromide, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 174120050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetrabutylammonium chloride | |
1112-67-0 | |
Authentic | |
Glass bottle | |
[CH3(CH2)3]4NCl | |
5 g | |
06,565; 07,353; 17,363 | |
Solubility in water: soluble. | |
CCCC[N+](CCCC)(CCCC)CCCC.[Cl-] | |
277.92 | |
CHEBI:51988 | |
95% | |
Crystalline Solid |
Also contains bromide | |
Brown to White | |
94% min. (on dry substance) (Argentometry) | |
C16H36ClN | |
MFCD00011635 | |
04, III, 292 | |
tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride | |
NHGXDBSUJJNIRV-UHFFFAOYSA-M | |
tetrabutylazanium;chloride | |
70681 | |
277.92 | |
Technical |
Chemical Identifiers
1112-67-0 | |
277.92 | |
NHGXDBSUJJNIRV-UHFFFAOYSA-M | |
70681 | |
tetrabutylazanium;chloride |
C16H36ClN | |
MFCD00011635 | |
tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride | |
CHEBI:51988 | |
CCCC[N+](CCCC)(CCCC)CCCC.[Cl-] |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 214-195-7
RUO – Research Use Only