Please login to your online account to display your discounted pricing

Tetrabutyl orthosilicate, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  313690050

 View more versions of this product

Catalog No. AC313690050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 320.54
Physical Form Liquid
Quantity 5g
Color Clear colorless
EINECS Number 225-305-8
Density 0.8990g/mL
TSCA TSCA
Packaging Glass bottle
Health Hazard 1 MOISTURE SENSITIVE
Refractive Index 1.4115 to 1.4145 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 95%
Chemical Name or Material Tetrabutyl orthosilicate
Linear Formula Si(OC4H9)4
Molecular Formula C16H36O4Si
MDL Number MFCD00009432
CAS 4766-57-8
Assay Percent Range 95%

Specifications

Chemical Name or Material Tetrabutyl orthosilicate
Name Note 95%
CAS 4766-57-8
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C16H36O4Si
Linear Formula Si(OC4H9)4
Formula Weight 320.54
MDL Number MFCD00009432
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents