Please login to your online account to display your discounted pricing

Tetrabutyl germanium, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrabutyl germanium
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H36Ge
Linear Formula [CH3(CH2)3]4Ge
Formula Weight 301.06
MDL Number MFCD00015224
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC321030050

  SDS
thermo fisher scientific
321030050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 301.06
Quantity 5g
Color Colorless to light yellow
EINECS Number 213-929-3
TSCA TSCA
Density 0.9300g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Chemical Name or Material Tetrabutyl germanium
Molecular Formula C16H36Ge
Linear Formula [CH3(CH2)3]4Ge
MDL Number MFCD00015224
CAS 1067-42-1
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.