Please login to your online account to display your discounted pricing

Tetrabromothiophene, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrabromothiophene
Name Note 99+%
CAS 3958-03-0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C4Br4S
Formula Weight 399.71
MDL Number MFCD00005419
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC137960250

  SDS
thermo fisher scientific
137960250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 399.71
Physical Form Crystalline Powder
Quantity 25g
Color Cream to brown
EINECS Number 223-554-7
Melting Point 112° to 116°C
Beilstein 17, 34
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Name Note 99+%
Chemical Name or Material Tetrabromothiophene
Molecular Formula C4Br4S
MDL Number MFCD00005419
CAS 3958-03-0
Assay Percent Range 99+%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.