missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabromohydroquinone, TCI America™

Manufacturer:  TCI America T179025G

 View more versions of this product

Catalog No. T179025G



Specifications

Specifications

Tetrabromohydroquinone
25g
C6H2Br4O2
tetrabromohydroquinone, 1,4-dihydroxytetrabromobenzene, tetrabromoquinol, 2,3,5,6-tetrabromohydroquinone, tetrabromohydroquinone, 2,3,5,6-tetrabromo-1,4-benzenediol, 1,2-benzenediol,3,4,5,6-tetrabromo, 1,4-benzenediol, 2,3,5,6-tetrabromo, 1,2,4,5-tetrabromo-3,6-dihydroxybenzene
C1(=C(C(=C(C(=C1Br)Br)O)Br)Br)O
425.696
425.70
≥97.0% (GC)
Undesignated
2641-89-6
MFCD00013970
DTFQULSULHRJOA-UHFFFAOYSA-N
2,3,5,6-tetrabromobenzene-1,4-diol
75840
Crystal-Powder at 20°C

Chemical Identifiers

2641-89-6
425.696
DTFQULSULHRJOA-UHFFFAOYSA-N
75840
C1(=C(C(=C(C(=C1Br)Br)O)Br)Br)O
C6H2Br4O2
MFCD00013970
tetrabromohydroquinone, 1,4-dihydroxytetrabromobenzene, tetrabromoquinol, 2,3,5,6-tetrabromohydroquinone, tetrabromohydroquinone, 2,3,5,6-tetrabromo-1,4-benzenediol, 1,2-benzenediol,3,4,5,6-tetrabromo, 1,4-benzenediol, 2,3,5,6-tetrabromo, 1,2,4,5-tetrabromo-3,6-dihydroxybenzene
2,3,5,6-tetrabromobenzene-1,4-diol
Safety and Handling

Safety and Handling

TSCA : No

Documents