missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetrabromocatechol, 96%

Manufacturer:  Alfa Aesar™ L0069014

 View more versions of this product

Catalog No. AAL0069014



Description

Description

Applications
Tetrabromocatechol is used as a pharmaceutical intermediate.

Solubility
It is partly miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases. Stable under recommended storage conditions.
Specifications

Specifications

Tetrabromocatechol
25g
C6H2Br4O2
2051548
It is partly miscible with water.
C1(=C(C(=C(C(=C1Br)Br)Br)Br)O)O
425.696
425.7
189°C to 193°C
488-47-1
MFCD00002189
1, 3,4,5,6-tetrabromo, 1,2-benzenediol, 3,4,5,6-tetrabromo, 3,4,5,6-tetrabromocatechol, 4-06-00-05624 beilstein handbook reference, pyrocatechol, tetrabromo, tetrabromocatechol, tetrabromopyrocatechol, unii-n642cph23m
OAUWOBSDSJNJQP-UHFFFAOYSA-N
3,4,5,6-tetrabromobenzene-1,2-diol
61127
96%

Chemical Identifiers

488-47-1
425.696
OAUWOBSDSJNJQP-UHFFFAOYSA-N
61127
C1(=C(C(=C(C(=C1Br)Br)Br)Br)O)O
C6H2Br4O2
MFCD00002189
1, 3,4,5,6-tetrabromo, 1,2-benzenediol, 3,4,5,6-tetrabromo, 3,4,5,6-tetrabromocatechol, 4-06-00-05624 beilstein handbook reference, pyrocatechol, tetrabromo, tetrabromocatechol, tetrabromopyrocatechol, unii-n642cph23m
3,4,5,6-tetrabromobenzene-1,2-diol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-678-9

RTECSNumber : UX2430000

TSCA : No

Recommended Storage : Ambient temperatures

Documents