missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabromo-p-benzoquinone, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC386190010

 View more versions of this product

Catalog No. AC386190010


 

Specifications Description & Specifications

Specifications

≥94%
423.68
1g
C6Br4O2
Glass bottle
Tetrabromo-p-benzoquinone
C6Br4(=O)2
Crystalline Powder
290° to 297°C (dec.)
Yellow
MFCD00013785
Authentic
488-48-2
95%

Chemical Identifiers

95%
C6Br4(=O)2
MFCD00013785
≥94%
C6Br4O2
423.68
Tetrabromo-p-benzoquinone

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning