Please login to your online account to display your discounted pricing

Tetrabromo-p-benzoquinone, 95%, ACROS Organics™

$58.12 - $58.12

Chemical Identifiers

Chemical Name or Material Tetrabromo-p-benzoquinone
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6Br4O2
Linear Formula C6Br4(=O)2
Formula Weight 423.68
MDL Number MFCD00013785
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC386190010

  SDS
thermo fisher scientific
AC386190010
Glass bottle 1g Each for $58.12

AC386190050

  SDS
thermo fisher scientific
AC386190050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 423.68
Melting Point 290° to 297°C (dec.)
Quantity 1g
Color Yellow
Molecular Formula C6Br4O2
MDL Number MFCD00013785
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material Tetrabromo-p-benzoquinone
CAS 488-48-2
Linear Formula C6Br4(=O)2
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.