missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabenzyl pyrophosphate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L09083MD

 View more versions of this product

Catalog No. AAL09083MD



Description

Description

Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acid and bases and reducing agents.
Specifications

Specifications

Tetrabenzyl pyrophosphate
58°C to 64°C
MFCD00051941
UN3261
Moisture sensitive
Slightly soluble in water.
C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4
538.473
538.48
990-91-0
C28H28O7P2
250 mg
2068292
tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
NSBNXCZCLRBQTA-UHFFFAOYSA-N
dibenzyl bis(phenylmethoxy)phosphoryl phosphate
563183
98%

Chemical Identifiers

990-91-0
538.473
NSBNXCZCLRBQTA-UHFFFAOYSA-N
563183
C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4
C28H28O7P2
MFCD00051941
tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
dibenzyl bis(phenylmethoxy)phosphoryl phosphate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Store at -20°C; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only