missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraamylammonium Chloride, TCI America™

Manufacturer:  TCI America T143325G

 View more versions of this product

Catalog No. T143325G



Specifications

Specifications

Tetraamylammonium Chloride
25g
C20H44ClN
2811
SXAWRMKQZKPHNJ-UHFFFAOYSA-M
tetrapentylazanium;chloride
78667
Crystal-Powder at 20°C
White
4965-17-7
MFCD00011857
Tetrapentylammonium Chloride
CCCCC[N+](CCCCC)(CCCCC)CCCCC.[Cl-]
334.029
334.03
≥98.0% (T)

Chemical Identifiers

4965-17-7
334.029
SXAWRMKQZKPHNJ-UHFFFAOYSA-M
78667
CCCCC[N+](CCCCC)(CCCCC)CCCCC.[Cl-]
C20H44ClN
MFCD00011857
Tetrapentylammonium Chloride
tetrapentylazanium;chloride
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

RTECSNumber : RZ9275000

TSCA : Yes

Documents