Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraammineplatinum(II) tetrachloroplatinate(II)

Chemical Identifiers

Chemical Name or Material Tetraammineplatinum(II) tetrachloroplatinate(II)
CAS 13820-46-7
Formula Weight 600.12
MDL Number MFCD01459899
Synonym Magnus Green salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3944403

  SDS
alfa aesar™
3944403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 600.12
Chemical Name or Material Tetraammineplatinum(II) tetrachloroplatinate(II)
TSCA TSCA
Density 4.000g/mL
Physical Form Powder
MDL Number MFCD01459899
CAS 13820-46-7
Synonym Magnus Green salt
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H334-H318-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 320°C (dec.)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P284a-P261-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H334-H318-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.