Learn More
Tetraammineplatinum(II) nitrate, Premion™, 99.99% (metals basis), Pt 50% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04389701
Description
Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetraammineplatinum(II) nitrate | |
White to Yellow | |
Crystals | |
99.99% (metals basis) | |
MFCD00011624 | |
Soluble in water. | |
N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
387.22 | |
387.21 |
262°C (decomposition) | |
20634-12-2 | |
0.25 g | |
H12N6O6Pt | |
tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt | |
RBAKORNXYLGSJB-UHFFFAOYSA-N | |
platinum(2+) tetraamine dinitrate | |
146543 |
Chemical Identifiers
20634-12-2 | |
387.22 | |
RBAKORNXYLGSJB-UHFFFAOYSA-N | |
146543 |
H12N6O6Pt | |
MFCD00011624 | |
tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt | |
N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c
H272-H315-H317-H319-H335
DOTInformation : DOT Class: 5.1; Packaging Group: II
EINECSNumber : 243-929-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only