missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraammineplatinum(II) hydrogen phosphate solution, Pt 0.5% w/w (cont. Pt)

Manufacturer:  Alfa Aesar™ 44076NB

 View more versions of this product

Catalog No. AA44076NB



Specifications

Specifications

Tetraammineplatinum (II) hydrogen phosphate solution
Colorless
127733-98-6
[Pt(NH3)4]HPO4
azane;hydrogen phosphate;platinum 2+
Fully miscible with water.
Liquid
1.013 g/mL
5g
H13N4O4PPt
MFCD03788258
Pt 0.5% w/w (cont. Pt)
359.18

Chemical Identifiers

127733-98-6
H13N4O4PPt
MFCD03788258
azane;hydrogen phosphate;platinum 2+
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : No

Recommended Storage : Ambient temperatures

Documents