Learn More
Tetraammineplatinum(II) hydrogen phosphate solution, Pt 0.5% w/w (cont. Pt), Thermo Scientific Chemicals
$1689.00 - $1689.00
Chemical Identifiers
CAS | 127733-98-6 |
---|---|
Molecular Formula | H13N4O4PPt |
Molecular Weight (g/mol) | 359.19 |
MDL Number | MFCD03788258 |
InChI Key | LSDWMQXWIQMHNQ-UHFFFAOYSA-L |
Synonym | azane;hydrogen phosphate;platinum 2+, platinum 2+ tetraamine hydrogen phosphate, platinum 2+ hydrogen phosphate-ammonia 1/1/4, platinum 2+ ion tetraamine hydrogen phosphate |
IUPAC Name | platinum(2+) tetraamine hydrogen phosphate |
SMILES | N.N.N.N.[Pt++].OP([O-])([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA44076NB
|
Thermo Scientific Chemicals
044076NB |
5 g |
Each for $1,689.00
|
|
|||||
Description
Tetraammineplatinum(II) hydrogen phosphate general application in pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
127733-98-6 | |
359.19 | |
LSDWMQXWIQMHNQ-UHFFFAOYSA-L | |
platinum(2+) tetraamine hydrogen phosphate |
H13N4O4PPt | |
MFCD03788258 | |
azane;hydrogen phosphate;platinum 2+, platinum 2+ tetraamine hydrogen phosphate, platinum 2+ hydrogen phosphate-ammonia 1/1/4, platinum 2+ ion tetraamine hydrogen phosphate | |
N.N.N.N.[Pt++].OP([O-])([O-])=O |
Specifications
127733-98-6 | |
1.013 g/mL | |
[Pt(NH3)4]HPO4 | |
5 g | |
Fully miscible with water. | |
N.N.N.N.[Pt++].OP([O-])([O-])=O | |
359.19 | |
Pt 0.5% w/w (cont. Pt) | |
Tetraammineplatinum (II) hydrogen phosphate solution |
Colorless | |
H13N4O4PPt | |
MFCD03788258 | |
azane;hydrogen phosphate;platinum 2+, platinum 2+ tetraamine hydrogen phosphate, platinum 2+ hydrogen phosphate-ammonia 1/1/4, platinum 2+ ion tetraamine hydrogen phosphate | |
LSDWMQXWIQMHNQ-UHFFFAOYSA-L | |
platinum(2+) tetraamine hydrogen phosphate | |
359.18 | |
Liquid |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only