missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraammineplatinum(II) hydrogen carbonate

Manufacturer:  Alfa Aesar™ 4404203

 View more versions of this product

Catalog No. AA4404203



Specifications

Specifications

Tetraammineplatinum (II) hydrogen carbonate
White
1g
C2H16N4O6Pt
MFCD03788256
AGUMVCVDCJQCIQ-UHFFFAOYSA-N
385.24
2.7 g/mL at 4°C
236°C (decomposition)
123439-82-7
[Pt(NH3)4](HCO3)2
c2h14n4o6pt
387.26
Crystalline

Chemical Identifiers

123439-82-7
387.26
AGUMVCVDCJQCIQ-UHFFFAOYSA-N
C2H16N4O6Pt
MFCD03788256
c2h14n4o6pt
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

EINECSNumber : 426-730-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents