missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraammineplatinum(II) chloride monohydrate, Premion™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 1083603

 View more versions of this product

Catalog No. AA1083603



Specifications

Specifications

Tetraammineplatinum(II) chloride monohydrate
(metals basis)
MFCD00149947
GWBDNMCYHWRSOH-UHFFFAOYSA-L
azane;platinum(2+);dichloride;hydrate
21946612
Crystalline
Premion™
Hygroscopic
250°C (dec.)
13933-33-0
Cl2H14N4OPt
azane;platinum 2+ ;dichloride;hydrate
N.N.N.N.O.[Cl-].[Cl-].[Pt+2]
352.123
352.13 (334.11 Anhydrous)
99.995%
2.74g/mL
White through Cream
1g

Chemical Identifiers

13933-33-0
352.123
GWBDNMCYHWRSOH-UHFFFAOYSA-L
21946612
N.N.N.N.O.[Cl-].[Cl-].[Pt+2]
Cl2H14N4OPt
MFCD00149947
azane;platinum 2+ ;dichloride;hydrate
azane;platinum(2+);dichloride;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H290-H315-H319
May be corrosive to metals.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P406
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
Store in corrosive resistant container with a resistant inner liner.

Warning

TSCA : TSCA

Documents