missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraamminepalladium(II) sulfate, 99.9% (metals basis), Pd 37.4% min

Manufacturer:  Alfa Aesar™ 4495603

 View more versions of this product

Catalog No. AA4495603



Specifications

Specifications

Tetraamminepalladium (II) sulfate
13601-06-4
H12N4O4PdS
tetraamminepalladium ii sulfate, tetraamminepalladium ii sulfate, palladium 2+ tetraamine sulfate, palladium 2+ sulfate-ammonia 1/1/4, palladium 2+ ion tetraamine sulfate
WSPHQZJZSJENIO-UHFFFAOYSA-L
azane;palladium(2+);sulfate
18319394
Crystalline
Pd 37.4% min
1g
(metals basis)
MFCD09953444
Pd 37.4% min
N.N.N.N.[O-]S(=O)(=O)[O-].[Pd+2]
270.6
270.61
99.9%

Chemical Identifiers

13601-06-4
270.6
WSPHQZJZSJENIO-UHFFFAOYSA-L
18319394
N.N.N.N.[O-]S(=O)(=O)[O-].[Pd+2]
H12N4O4PdS
MFCD09953444
tetraamminepalladium ii sulfate, tetraamminepalladium ii sulfate, palladium 2+ tetraamine sulfate, palladium 2+ sulfate-ammonia 1/1/4, palladium 2+ ion tetraamine sulfate
azane;palladium(2+);sulfate
SDS Documents