missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraamminepalladium(II) hydrogen carbonate

Manufacturer:  Alfa Aesar™ 4508206

 View more versions of this product

Catalog No. AA4508206



Specifications

Specifications

Tetraamminepalladium (II) hydrogen carbonate
Colorless to Yellow
134620-00-1
[Pd(NH3)4](HCO3)2
UN3077
QYYPTZGPXKUKRM-UHFFFAOYSA-L
azane;hydrogen carbonate;palladium(2+)
153931
Crystalline
2.04g/mL
5g
C2H14N4O6Pd
MFCD09953449
palladium 2+ , tetraammine-, sp-4-1-, carbonate 1:2, azane; hydrogen carbonate; palladium 2+, tetraamminepalladium ii hydrogencarbonate, ammonia; hydrogen carbonate; palladium 2+, palladium 2+ tetraamine dibicarbonate, tetraammine palladium ii hydrogen carbonate, palladium 2+ ion tetraamine dibicarbonate
C(=O)(O)[O-].C(=O)(O)[O-].N.N.N.N.[Pd+2]
296.576
296.58

Chemical Identifiers

134620-00-1
296.576
QYYPTZGPXKUKRM-UHFFFAOYSA-L
153931
C(=O)(O)[O-].C(=O)(O)[O-].N.N.N.N.[Pd+2]
C2H14N4O6Pd
MFCD09953449
palladium 2+ , tetraammine-, sp-4-1-, carbonate 1:2, azane; hydrogen carbonate; palladium 2+, tetraamminepalladium ii hydrogencarbonate, ammonia; hydrogen carbonate; palladium 2+, palladium 2+ tetraamine dibicarbonate, tetraammine palladium ii hydrogen carbonate, palladium 2+ ion tetraamine dibicarbonate
azane;hydrogen carbonate;palladium(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H318-H371-H373-H302-H317
Causes serious eye damage.
May cause damage to organs.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P273-P305+P351+P338-P309+P311-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : RT3597200

TSCA : TSCA

SDS Documents