missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraamminepalladium(II) chloride monohydrate, 99.9% (metals basis), Pd 39% min

$210.24 - $1,001.24

Chemical Identifiers

CAS 13933-31-8
Molecular Formula Cl2H14N4OPd
Molecular Weight (g/mol) 263.46
MDL Number MFCD00151033
InChI Key WVCXSPJPERKPJS-UHFFFAOYSA-L
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1103604
View Documents
Alfa Aesar™
1103604
2g
Each for $210.24
 
AA1103609
View Documents
Alfa Aesar™
1103609
10g
Each for $1,001.24
 
Description

Description

Specifications

Chemical Identifiers

13933-31-8
263.46
WVCXSPJPERKPJS-UHFFFAOYSA-L
Cl2H14N4OPd
MFCD00151033

Specifications

1.91 g/mL
Colorless to Yellow
2g
Cl2H14N4OPd
UN3077
Soluble in water.
263.46
Crystalline
Tetraamminepalladium(II) chloride monohydrate
Hygroscopic
120°C (decomposition)
13933-31-8
MFCD00151033
Pd 39% min.
WVCXSPJPERKPJS-UHFFFAOYSA-L
263.45 (245.43 Anhydrous)
99.9%
(metals basis)
Safety and Handling

Safety and Handling

GHS H Statement
H290-H302-H317-H319
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.

P234-P261-P264b-P270-P280i-P285-P301+P312-P304+P341-P305+P351+P338-P330-P342+P311-P390-P501c

H290-H302-H319-H334

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS