missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraamminepalladium(II) chloride monohydrate, 99.9% (metals basis), Pd 39% min

Manufacturer:  Alfa Aesar™ 1103604

 View more versions of this product

Catalog No. AA1103604



Specifications

Specifications

Tetraamminepalladium(II) chloride monohydrate
Hygroscopic
120°C (decomposition)
13933-31-8
Cl2H14N4OPd
UN3077
Soluble in water.
263.46
Crystalline
1.91 g/mL
Colorless to Yellow
2g
(metals basis)
MFCD00151033
Pd 39% min.
WVCXSPJPERKPJS-UHFFFAOYSA-L
263.45 (245.43 Anhydrous)
99.9%

Chemical Identifiers

13933-31-8
263.46
WVCXSPJPERKPJS-UHFFFAOYSA-L
Cl2H14N4OPd
MFCD00151033
Safety and Handling

Safety and Handling

GHS H Statement
H290-H302-H317-H319
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.

P234-P261-P264b-P270-P280i-P285-P301+P312-P304+P341-P305+P351+P338-P330-P342+P311-P390-P501c

H290-H302-H319-H334

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents