Please login to your online account to display your discounted pricing

Tetraallyltin, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetraallyltin
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H20Sn
Linear Formula (CH2=CHCH2)4Sn
Formula Weight 282.98
MDL Number MFCD00008637
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382490010

  SDS
thermo fisher scientific
AC382490010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC382490050

  SDS
thermo fisher scientific
AC382490050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 282.98
Physical Form Liquid
Quantity 1g
Color Clear colorless
Density 1.1790g/mL
Infrared Spectrum Authentic
Chemical Name or Material Tetraallyltin
CAS 7393-43-3
MDL Number MFCD00008637
Molecular Formula C12H20Sn
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5400 to 1.5420 (20°C, 589nm)
Linear Formula (CH2=CHCH2)4Sn
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger