Please login to your online account to display your discounted pricing

Tetraallyltin, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC382490050

 View more versions of this product

Catalog No. AC382490050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 282.98
Quantity 5g
Color Clear colorless
Density 1.1790g/mL
Refractive Index 1.5400 to 1.5420 (20°C, 589nm)
Health Hazard 1 Danger
Chemical Name or Material Tetraallyltin
MDL Number MFCD00008637
Assay Percent Range 97%
CAS 7393-43-3
Molecular Formula C12H20Sn
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH2=CHCH2)4Sn
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetraallyltin
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H20Sn
Linear Formula (CH2=CHCH2)4Sn
Formula Weight 282.98
MDL Number MFCD00008637
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

SDS & Safety Documents