missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetra-O-acetyl-beta-D-ribofuranose 98.0+%, TCI America™

Manufacturer:  Tci America R006625G

 View more versions of this product

Catalog No. R006625G



Specifications

Specifications

Tetra-O-acetyl-beta-D-ribofuranose
83°C
13035-61-5
MFCD00005358
IHNHAHWGVLXCCI-FDYHWXHSSA-N
[(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
83064
Crystal-Powder at 20°C
White
25g
C13H18O9
beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
318.278
318.28
≥98.0% (GC)

Chemical Identifiers

13035-61-5
318.278
IHNHAHWGVLXCCI-FDYHWXHSSA-N
83064
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
C13H18O9
MFCD00005358
beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
[(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
Safety and Handling

Safety and Handling

TSCA : No

Documents