Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-propyltin

Chemical Identifiers

Chemical Name or Material Tetra-n-propyltin
CAS 2176-98-9
Molecular Formula C12H28Sn
Linear Formula (CH3CH2CH2)4Sn
Formula Weight 291.04
MDL Number MFCD00015196
UN Number UN2788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA7113709

  SDS
alfa aesar™
7113709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 291.04
Molecular Formula C12H28Sn
Health Hazard 1 Danger
Odor Unpleasant
Boiling Point 222°C
MDL Number MFCD00015196
Linear Formula (CH3CH2CH2)4Sn
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Health Hazard 2 GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2788
Density 1.11g/mL
Chemical Name or Material Tetra-n-propyltin
Quantity 10g
Physical Form Liquid
CAS 2176-98-9
Color Colorless
Refractive Index 1.4745
RTECS Number WH8636500
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.