Learn More
Tetra-n-propylammonium perruthenate(VII), 98%, Thermo Scientific Chemicals
$139.81 - $139.81
Chemical Identifiers
CAS | 114615-82-6 |
---|---|
Molecular Formula | C12H35NO4Ru |
Molecular Weight (g/mol) | 358.485 |
MDL Number | MFCD00074914 |
InChI Key | HUCLFLGLPCVDMZ-UHFFFAOYSA-M |
Synonym | 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1- |
PubChem CID | 127020979 |
IUPAC Name | ruthenium;tetrapropylazanium;hydroxide;trihydrate |
SMILES | CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2451102
|
Thermo Scientific Chemicals
B2451102 |
0.5 g |
Each for $139.81
|
|
|||||
Description
Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.
Solubility
Insoluble in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, reducing agents, heat, organic materials, metal powders, heat.
Chemical Identifiers
114615-82-6 | |
358.485 | |
HUCLFLGLPCVDMZ-UHFFFAOYSA-M | |
127020979 | |
CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru] |
C12H35NO4Ru | |
MFCD00074914 | |
1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1- | |
ruthenium;tetrapropylazanium;hydroxide;trihydrate |
Specifications
160°C (decomposition) | |
0.5 g | |
(CH3(CH2)2)4NRuO4 | |
UN1479 | |
Insoluble in water. | |
CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru] | |
358.485 | |
351.43 | |
Hygroscopic |
114615-82-6 | |
C12H35NO4Ru | |
MFCD00074914 | |
1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1- | |
HUCLFLGLPCVDMZ-UHFFFAOYSA-M | |
ruthenium;tetrapropylazanium;hydroxide;trihydrate | |
127020979 | |
98% | |
Tetra-n-propylammonium perruthenate(VII) |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335-H500
DOTInformation : Hazard Class: 5.1; Packaging Group: II
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only