Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-propylammonium iodide, 98+%

$64.93 - $64.93

Chemical Identifiers

Chemical Name or Material Tetra-n-propylammonium iodide
CAS 631-40-3
Molecular Formula (CH3CH2CH2)4NI
Formula Weight 313.27
MDL Number MFCD00011842
Beilstein 3567386
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1596114

  SDS
alfa aesar™
A1596114
25g Each for $64.93
Description & Specifications

Specifications

Formula Weight 313.27
Chemical Name or Material Tetra-n-propylammonium iodide
TSCA TSCA
Assay ≥98%
Molecular Formula (CH3CH2CH2)4NI
MDL Number MFCD00011842
CAS 631-40-3
Beilstein 3567386
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 283°C (dec.)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.