Learn More
Tetra-n-propylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01745636
Description
Tetra-n-propylammonium hydroxide is used as an intermediate, ion exchange agent, surface-active agent and solvent. Further, it is used as an antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It finds application as curing accelerators. In addition to this, it acts as a phase transfer catalyst in organic synthesis to enhance the reaction rate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetra-n-propylammonium hydroxide | |
Colorless to Yellow | |
102°C | |
1.3716 | |
MFCD00009360 | |
UN3267 | |
Air Sensitive | |
Miscible with strong electrolytes. | |
[OH-].CCC[N+](CCC)(CCC)CCC | |
203.37 | |
203.37 | |
Liquid |
4499-86-9 | |
0.99 g/mL | |
C12H29NO | |
(CH3CH2CH2)4NOH | |
500 g | |
4207983 | |
tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide | |
LPSKDVINWQNWFE-UHFFFAOYSA-M | |
tetrapropylazanium;hydroxide | |
20586 | |
40% w/w aq. soln. |
Chemical Identifiers
4499-86-9 | |
203.37 | |
LPSKDVINWQNWFE-UHFFFAOYSA-M | |
20586 | |
[OH-].CCC[N+](CCC)(CCC)CCC |
C12H29NO | |
MFCD00009360 | |
tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide | |
tetrapropylazanium;hydroxide |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 224-800-6
RTECSNumber : BS8400000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only