missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-propylammonium hydrogen sulfate, 98%

Manufacturer:  Alfa Aesar™ A1200203

 View more versions of this product

Catalog No. AAA1200203



Specifications

Specifications

Tetra-n-propylammonium hydrogen sulfate
98%
(CH3CH2CH2)4NHSO4
MOXJKKOSZCHGEU-UHFFFAOYSA-M
hydrogen sulfate;tetrapropylazanium
16218669
158°C to 160°C
56211-70-2
C12H29NO4S
MFCD00038764
CCC[N+](CCC)(CCC)CCC.OS(=O)(=O)[O-]
283.427
283.43
1g

Chemical Identifiers

56211-70-2
283.427
MOXJKKOSZCHGEU-UHFFFAOYSA-M
hydrogen sulfate;tetrapropylazanium
C12H29NO4S
MFCD00038764
16218669
CCC[N+](CCC)(CCC)CCC.OS(=O)(=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents