missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-propylammonium chloride, 99+%

Manufacturer:  Alfa Aesar™ 1937914

 View more versions of this product

Catalog No. AA1937914



Specifications

Specifications

Tetra-n-propylammonium chloride
Hygroscopic
240°C to 242°C
5810-42-4
(CH3CH2CH2)4NCl
tetrapropylammonium chloride, tetrapropyl ammonium chloride, tetra-n-propylammonium chloride, 1-propanaminium, n,n,n-tripropyl-, chloride, 1-propanaminium, n,n,n-tripropyl-, chloride 1:1, n,n,n-tripropylpropan-1-aminium chloride, tetrapropylazanium chloride, acmc-209m2k, ksc491c1h, tetra-n-propylammonium chloride 1g
FBEVECUEMUUFKM-UHFFFAOYSA-M
tetrapropylazanium;chloride
79880
Crystalline
Odorless
White
25g
C12H28ClN
MFCD00038729
Soluble in water, acetone
CCC[N+](CCC)(CCC)CCC.[Cl-]
221.813
221.87
≥99%

Chemical Identifiers

5810-42-4
221.813
FBEVECUEMUUFKM-UHFFFAOYSA-M
79880
CCC[N+](CCC)(CCC)CCC.[Cl-]
C12H28ClN
MFCD00038729
tetrapropylammonium chloride, tetrapropyl ammonium chloride, tetra-n-propylammonium chloride, 1-propanaminium, n,n,n-tripropyl-, chloride, 1-propanaminium, n,n,n-tripropyl-, chloride 1:1, n,n,n-tripropylpropan-1-aminium chloride, tetrapropylazanium chloride, acmc-209m2k, ksc491c1h, tetra-n-propylammonium chloride 1g
tetrapropylazanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents