Learn More
Tetra-n-propylammonium chloride, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01937906
Description
Tetra-n-propylammonium chloride is used as phase-transfer type applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetra-n-propylammonium chloride | |
240°C to 242°C | |
Odorless | |
(CH3CH2CH2)4NCl | |
5 g | |
tetrapropylammonium chloride, tetrapropyl ammonium chloride, tetra-n-propylammonium chloride, 1-propanaminium, n,n,n-tripropyl-, chloride, 1-propanaminium, n,n,n-tripropyl-, chloride 1:1, n,n,n-tripropylpropan-1-aminium chloride, tetrapropylazanium chloride, acmc-209m2k, ksc491c1h, tetra-n-propylammonium chloride 1g | |
FBEVECUEMUUFKM-UHFFFAOYSA-M | |
tetrapropylazanium;chloride | |
79880 | |
≥99% |
5810-42-4 | |
White | |
C12H28ClN | |
MFCD00038729 | |
Hygroscopic | |
Soluble in water, acetone | |
CCC[N+](CCC)(CCC)CCC.[Cl-] | |
221.813 | |
221.87 | |
Crystalline |
Chemical Identifiers
5810-42-4 | |
221.813 | |
FBEVECUEMUUFKM-UHFFFAOYSA-M | |
79880 | |
CCC[N+](CCC)(CCC)CCC.[Cl-] |
C12H28ClN | |
MFCD00038729 | |
tetrapropylammonium chloride, tetrapropyl ammonium chloride, tetra-n-propylammonium chloride, 1-propanaminium, n,n,n-tripropyl-, chloride, 1-propanaminium, n,n,n-tripropyl-, chloride 1:1, n,n,n-tripropylpropan-1-aminium chloride, tetrapropylazanium chloride, acmc-209m2k, ksc491c1h, tetra-n-propylammonium chloride 1g | |
tetrapropylazanium;chloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 227-375-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only