missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetra-n-hexylammonium perchlorate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03083006

 View more versions of this product

Catalog No. AA3083006



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetra-n-hexylammonium perchlorate
C24H52ClNO4
5 g
tetra-n-hexylammonium perchlorate, tetrahexylammonium perchlorate, tetrahexylazanium perchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate, n,n,n-trihexylhexan-1-aminiumperchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate 1:1, tetra-n-hexyl ammonium perchlorate
[O-][Cl](=O)(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
454.13
454.13
4656-81-9
MFCD00043185
UN1479
RLCKPGXHGKSGOS-UHFFFAOYSA-M
tetrahexylazanium;perchlorate
78388
Crystalline

Chemical Identifiers

4656-81-9
454.13
RLCKPGXHGKSGOS-UHFFFAOYSA-M
78388
C24H52ClNO4
MFCD00043185
tetra-n-hexylammonium perchlorate, tetrahexylammonium perchlorate, tetrahexylazanium perchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate, n,n,n-trihexylhexan-1-aminiumperchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate 1:1, tetra-n-hexyl ammonium perchlorate
[O-][Cl](=O)(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 225-095-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only