Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetra-n-hexylammonium benzoate, ca 85%, water ca 10%

$46.80 - $549.53

Chemical Identifiers

Chemical Name or Material Tetra-n-hexylammonium benzoate
CAS 16436-29-6
Molecular Formula [CH3(CH2)5]4N[O2C(C6H5)]
Formula Weight 456.73
MDL Number MFCD00041982
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2529103

 
alfa aesar™
B2529103
1g Each for $46.80

AAB2529106

 
alfa aesar™
B2529106
5g Each for $153.93

AAB2529114

 
alfa aesar™
B2529114
25g Each for $549.53
Description & Specifications

Specifications

Formula Weight 456.73
Chemical Name or Material Tetra-n-hexylammonium benzoate
TSCA TSCA
Assay ≈85%
Assay Percent Notes water ≈10%
Molecular Formula [CH3(CH2)5]4N[O2C(C6H5)]
MDL Number MFCD00041982
CAS 16436-29-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.